Search by lot number or partial lot number
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile. Not available in all geographic regions.
Product Type | Base Tube |
Description | Round Base Plastic Tubes |
Material | Polystyrene |
Diameter (Metric) Outer | 16 mm |
Packaging Quantity | 1500 |
Product Line | Sterilin™ |
Sterility | Non-sterile |
Tubing Size | 11.0 mL |
Unit Size | Case of 750 |
Product Type | Base Tube |
Description | Round Base Plastic Tubes |
Material | Polystyrene |
Diameter (Metric) Outer | 6 mm |
Packaging Quantity | 1000 |
Product Line | Sterilin™ |
Sterility | Non-sterile |
Tubing Size | 0.6 mL |
Unit Size | Each |
Description | Round Base Plastic Tubes |
Material | Polystyrene |
Diameter (Metric) Outer | 6 mm |
Packaging Quantity | 1000 |
Sterility | Non-sterile |
Tubing Size | 1.0 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Tubing Size | Packaging Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
30932 | Case of 750 | 11.0 mL | 1500 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
RT15 | Each | 0.6 mL | 1000 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
RT20 | Each | 1.0 mL | 1000 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|