Thermo Scientific Chemicals

3-Iodo-o-xylene, 98%, Thermo Scientific Chemicals

Catalog number: 309350050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Iodo-o-xylene, 98%, Thermo Scientific Chemicals

Catalog number: 309350050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 309350050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
31599-60-7
IUPAC Name
1-iodo-2,3-dimethylbenzene
Molecular Formula
C8H9I
InChI Key
DANMWBNOPFBJSZ-UHFFFAOYSA-N
SMILES
CC1=CC=CC(I)=C1C
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.6060 to 1.6080 (20°C, 589 nm)
Appearance (Color)
Clear red-brown to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text