4-Methoxyphenylboronic acid, 97%
4-Methoxyphenylboronic acid, 97%
4-Methoxyphenylboronic acid, 97%
4-Methoxyphenylboronic acid, 97%
Thermo Scientific Chemicals

4-Methoxyphenylboronic acid, 97%

CAS: 5720-07-0 | C7H9BO3 | 151.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 309480010
Price (EUR)
37,91
Online Exclusive
44,60
Save 6,69 (15%)
Each
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,91
Online Exclusive
44,60
Save 6,69 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name(4-methoxyphenyl)boronic acid
Molecular FormulaC7H9BO3
InChI KeyVOAAEKKFGLPLLU-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol)151.96
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Titration with NaOH>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link