Thermo Scientific Chemicals

D-Trehalose, 99%, anhydrous

Catalog number: 309870250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

D-Trehalose, 99%, anhydrous

Catalog number: 309870250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 309870250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
99-20-7
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
HDTRYLNUVZCQOY-LBYRQFQKNA-N
SMILES
OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Appearance (Color)
White to almost white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=98.5 %
Water
=<1.0 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text