D-Glucuronic acid, 98%, Thermo Scientific Chemicals
D-Glucuronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Glucuronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
Packaging:
Plastic bottle
Catalog number 309880250
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
Add to cart
D-Glucuronic acid, 98%, Thermo Scientific Chemicals
Catalog number309880250
Price (USD)/ Each
152.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
179.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
CAS6556-12-3
IUPAC Name(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
Molecular FormulaC6H10O7
InChI KeyIAJILQKETJEXLJ-QTBDOELSSA-N
SMILESO[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Appearance (Color)White to off-white
Infrared spectrumConforms
Specific optical rotationafter 4 h
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only