Thermo Scientific Chemicals

D-Glucuronic acid, 98%, Thermo Scientific Chemicals

Catalog number: 309881000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

D-Glucuronic acid, 98%, Thermo Scientific Chemicals

Catalog number: 309881000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 309881000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6556-12-3
IUPAC Name
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
Molecular Formula
C6H10O7
InChI Key
IAJILQKETJEXLJ-QTBDOELSSA-N
SMILES
O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C(O)=O
Water
=<1 % (K.F.)
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Specific optical rotation
after 4 h
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text