Thermo Scientific Chemicals

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%, Thermo Scientific Chemicals

Catalog number: 310060250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%, Thermo Scientific Chemicals

Catalog number: 310060250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 310060250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
68641-49-6
IUPAC Name
bis(2-oxo-1,3-oxazolidin-3-yl)phosphinoyl chloride
Molecular Formula
C6H8ClN2O5P
InChI Key
KLDLRDSRCMJKGM-UHFFFAOYSA-N
SMILES
ClP(=O)(N1CCOC1=O)N1CCOC1=O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Titration Argentometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text