Informational:Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual. Compare options for filtered tips »
Thermo Scientific™
ART™ Non-Filtered Standard Hinged Rack Pipette Tips
Access tips with one hand using Thermo ScientificTM ARTTM Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.
Catalog Number
3101-HR
Unit Size
Case of 3072
Description
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack
Volume (Metric)
1000 μL
Sterility
Sterile
Fit for Purpose
Sterile
Price (USD)
399.68 Online price
471.00
Availability
-
Full specifications
Quantity
Description
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack
Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
Sterile tips are DNase, RNase, DNA, ATP, and Endotoxin Free
Racks offer quick and easy access to pipette tips, saving valuable time
Front latch closes and locks, ensuring safe and secure storage throughout