Dimethylphenylphosphine, 96%, Thermo Scientific Chemicals
Dimethylphenylphosphine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylphenylphosphine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 310100250
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
469.00 
Save 70.35 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
469.00 
Save 70.35 (15%)
Add to cart
Dimethylphenylphosphine, 96%, Thermo Scientific Chemicals
Catalog number310100250
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
469.00 
Save 70.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedimethyl(phenyl)phosphane
Molecular FormulaC8H11P
InChI KeyHASCQPSFPAKVEK-UHFFFAOYSA-N
SMILESCP(C)C1=CC=CC=C1
Molecular Weight (g/mol)138.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=95 %
Appearance (Color)Clear colorless
Refractive index1.5610 to 1.5660 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
1x1 image pixel for data collection