Thermo Scientific Chemicals

Diphenylphosphine oxide, 97%, Thermo Scientific Chemicals

Catalog number: 310120010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenylphosphine oxide, 97%, Thermo Scientific Chemicals

Catalog number: 310120010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 310120010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4559-70-0
IUPAC Name
(phenylphosphoroso)benzene
Molecular Formula
C12H11OP
InChI Key
ASUOLLHGALPRFK-UHFFFAOYSA-N
SMILES
O=P(C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Color)
White to yellow
Appearance (Form)
Powder or crystals
Melting point
50°C to 57°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text