Diphenylphosphine oxide, 97%, Thermo Scientific Chemicals
Diphenylphosphine oxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylphosphine oxide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 310120050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4559-70-0
IUPAC Name(phenylphosphoroso)benzene
Molecular FormulaC12H11OP
InChI KeyASUOLLHGALPRFK-UHFFFAOYSA-N
SMILESO=P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)White to yellow
Appearance (Form)Powder or crystals
Melting point50°C to 57°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only