Ethyl isobutyrate, 99%, Thermo Scientific Chemicals
Ethyl isobutyrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl isobutyrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 310515000
Price (USD)/ Each
46.10
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
46.10
Add to cart
Ethyl isobutyrate, 99%, Thermo Scientific Chemicals
Catalog number310515000
Price (USD)/ Each
46.10
-
Add to cart
Chemical Identifiers
CAS97-62-1
IUPAC Nameethyl 2-methylpropanoate
Molecular FormulaC6H12O2
InChI KeyWDAXFOBOLVPGLV-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only