Octyl acetate, 99+%, Thermo Scientific Chemicals
Octyl acetate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Octyl acetate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 310631000
Price (USD)/ Each
25.60
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
25.60
Add to cart
Octyl acetate, 99+%, Thermo Scientific Chemicals
Catalog number310631000
Price (USD)/ Each
25.60
-
Add to cart
Chemical Identifiers
CAS112-14-1
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=99.0 %
Refractive index1.4175 to 1.4195 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only