Thermo Scientific Chemicals

Ammonium tetrafluoroborate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 310960025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium tetrafluoroborate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 310960025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 310960025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13826-83-0
IUPAC Name
ammonium tetrafluoroboranuide
Molecular Formula
BF4H4N
InChI Key
PDTKOBRZPAIMRD-UHFFFAOYSA-O
SMILES
[NH4+].F[B-](F)(F)F
Appearance (Color)
White
Titration with NaOH
>=98.5 %
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text