1,2-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 311350010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,2-dibromo-3,4,5,6-tetrafluorobenzene
Molecular FormulaC6Br2F4
InChI KeyIPLUWQPTPKNBRD-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(Br)C(Br)=C1F
Molecular Weight (g/mol)307.87
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5145 to 1.5175 (20°C, 589 nm)
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only