3,5-Difluoroaniline, 98%, Thermo Scientific Chemicals
3,5-Difluoroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Difluoroaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 311420010
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.00 
Save 4.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.00 
Save 4.35 (15%)
Add to cart
3,5-Difluoroaniline, 98%, Thermo Scientific Chemicals
Catalog number311420010
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.00 
Save 4.35 (15%)
-
Add to cart
Chemical Identifiers
CAS372-39-4
IUPAC Name3,5-difluoroaniline
Molecular FormulaC6H5F2N
InChI KeyKQOIBXZRCYFZSO-UHFFFAOYSA-N
SMILESNC1=CC(F)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Low melting solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only