Thermo Scientific Chemicals

3,5-Difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 311420100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 311420100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 311420100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
372-39-4
IUPAC Name
3,5-difluoroaniline
Molecular Formula
C6H5F2N
InChI Key
KQOIBXZRCYFZSO-UHFFFAOYSA-N
SMILES
NC1=CC(F)=CC(F)=C1
Appearance
May darken on storage
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text