Thermo Scientific Chemicals

2,6-Difluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 311630050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Difluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 311630050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 311630050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28177-48-2
IUPAC Name
2,6-difluorophenol
Molecular Formula
C6H4F2O
InChI Key
CKKOVFGIBXCEIJ-UHFFFAOYSA-N
SMILES
OC1=C(F)C=CC=C1F
Appearance (Color)
Colorless
Appearance (Form)
Low melting crystalline solid
Melting point
38°C to 42°C
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text