5-Fluoroindole, 98%, Thermo Scientific Chemicals
5-Fluoroindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoroindole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 311920010
Price (USD)/ Each
49.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.10 
Save 8.45 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.10 
Save 8.45 (15%)
Add to cart
5-Fluoroindole, 98%, Thermo Scientific Chemicals
Catalog number311920010
Price (USD)/ Each
49.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.10 
Save 8.45 (15%)
-
Add to cart
Chemical Identifiers
CAS399-52-0
IUPAC Name5-fluoro-1H-indole
Molecular FormulaC8H6FN
InChI KeyODFFPRGJZRXNHZ-UHFFFAOYSA-N
SMILESFC1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point44°C to 48°C
Appearance (Color)White to beige
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only