Thermo Scientific™

Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack

Catalog number: 312002-9025
Have Questions?
Thermo Scientific™

Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack

Catalog number: 312002-9025
Have Questions?

Sample, store and ship small-volume, valuable reagents. Thermo Scientific™ Nalgene Diagnostic Bottles with closures are used in test kits and many small-volume applications. Natural, translucent high-density polyethylene provides a durable design with uniform walls that are resistant to splitting or puncturing.

 
Catalog Number
312002-9025
Unit Size
Case of 2000
Capacity (Metric)
8 mL
Neck Finish
20-415 mm
Price (USD)
Availability
-
Full specifications
Quantity
Capacity0.25 oz., 8 mL
Capacity (Metric)8 mL
Capacity (English)0.25 oz.
ColorTranslucent
Diameter (Metric) Inner Neck14 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish20-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthNarrow
Height (Metric)45 mm
Product TypeSample Container
Height (English)1.77 in.
Closure LinerUnlined
Diameter (Metric)25 mm
Through the Closure DispensingNo
Unit SizeCase of 2000
Catalog Number
Unit Size
Case of 2000
Capacity (Metric)
4 mL
Neck Finish
13-415 mm
Price (USD)
Availability
-
Full specifications
Quantity
Capacity0.125 oz., 4 mL
Capacity (Metric)4 mL
Capacity (English)0.125 oz.
ColorTranslucent
Diameter (Metric) Inner Neck8 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish13-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthNarrow
Height (Metric)41 mm
Product TypeSample Container
Height (English)1.61 in.
Closure LinerUnlined
Diameter (Metric)16 mm
Through the Closure DispensingNo
Unit SizeCase of 2000
Catalog Number
Unit Size
Case of 2000
Capacity (Metric)
15 mL
Neck Finish
20-415 mm
Price (USD)
Availability
-
Full specifications
Quantity
Capacity0.5 oz., 15 mL
Capacity (Metric)15 mL
Capacity (English)0.5 oz.
ColorNatural
Diameter (Metric) Inner Neck14 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish20-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)58 mm
Product TypeSample Container
Width (English)0.98 in.
Height (English)2.3 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size20-415 mm
Diameter (Metric)25 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCase of 2000
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeCapacity (Metric)Neck FinishPrice (USD)AvailabilityQuantity
312002-9025Full specifications
Case of 20008 mL20-415 mm
-
Capacity0.25 oz., 8 mL
Capacity (Metric)8 mL
Capacity (English)0.25 oz.
ColorTranslucent
Diameter (Metric) Inner Neck14 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish20-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthNarrow
Height (Metric)45 mm
Product TypeSample Container
Height (English)1.77 in.
Closure LinerUnlined
Diameter (Metric)25 mm
Through the Closure DispensingNo
Unit SizeCase of 2000
312002-9125Full specifications
Case of 20004 mL13-415 mm
-
Capacity0.125 oz., 4 mL
Capacity (Metric)4 mL
Capacity (English)0.125 oz.
ColorTranslucent
Diameter (Metric) Inner Neck8 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish13-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthNarrow
Height (Metric)41 mm
Product TypeSample Container
Height (English)1.61 in.
Closure LinerUnlined
Diameter (Metric)16 mm
Through the Closure DispensingNo
Unit SizeCase of 2000
312002-9050Full specifications
Case of 200015 mL20-415 mm
-
Capacity0.5 oz., 15 mL
Capacity (Metric)15 mL
Capacity (English)0.5 oz.
ColorNatural
Diameter (Metric) Inner Neck14 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish20-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)58 mm
Product TypeSample Container
Width (English)0.98 in.
Height (English)2.3 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size20-415 mm
Diameter (Metric)25 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCase of 2000
Showing 3 of 3
Features:
  • Nonsterile
  • Natural, linerless polypropylene closures
  • Durable, uniform walls resistant to splitting or puncturing
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton. Packed 2000 per case.

.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text