Thermo Scientific Chemicals

1-Fluoropyridinium tetrafluoroborate, 95+%, Thermo Scientific Chemicals

Catalog number: 312010010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Fluoropyridinium tetrafluoroborate, 95+%, Thermo Scientific Chemicals

Catalog number: 312010010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 312010010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107264-09-5
IUPAC Name
1-fluoropyridin-1-ium
Molecular Formula
C5H5FN
InChI Key
UWDCUCCPBLHLTI-UHFFFAOYSA-N
SMILES
F[N+]1=CC=CC=C1
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Titration with NaOH
95.0 to 105.0 %
Appearance (Color)
White to light yellow to light beige
Melting point
190°C to 197°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text