Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store and ship liquid reagents with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure. Leakproof† when used with Nalgene closures and constructed of durable high-density polyethylene, these safe, economical bottles are recommended for those designing, assembling and certifying their own combination packaging.
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | White |
Description | IP2 Wide-Mouth Bottles |
Diameter (Metric) Inner Neck | 44 mm |
Diameter (Metric) Inner | 44 mm |
For Use With (Application) | Storage and Shipment of Liquid Reagents |
Height (Metric) With Closure | 168 mm |
Material | HDPE |
No. per Case | 125 |
No. per Pack | 125 |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. |
Closure Included | Yes |
Mouth | Wide |
Shape | Round |
Height (Metric) | 168 mm |
Product Type | IP2 Bottle |
Height (English) | 6.6 in. |
Closure Material | Polypropylene |
Closure Size | 53-415 mm |
Diameter (Metric) | 73 mm |
Unit Size | Case of 125 |
Capacity | 4 oz., 125 mL |
Capacity (Metric) | 125 mL |
Capacity (English) | 4 oz. |
Color | White |
Description | IP2 Wide-Mouth Bottles |
Diameter (Metric) Inner Neck | 28 mm |
Diameter (Metric) Inner | 28 mm |
For Use With (Application) | Storage and Shipment of Liquid Reagents |
Height (Metric) With Closure | 99 mm |
Material | HDPE |
No. per Case | 500 |
No. per Pack | 500 |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. |
Closure Included | Yes |
Mouth | Wide |
Shape | Round |
Height (Metric) | 99 mm |
Product Type | IP2 Bottle |
Height (English) | 3.8 in. |
Closure Material | Polypropylene |
Closure Size | 38-415 mm |
Diameter (Metric) | 50 mm |
Unit Size | Case of 500 |
Capacity | 8 oz., 250 mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8 oz. |
Color | White |
Description | IP2 Wide-Mouth Bottles |
Diameter (Metric) Inner Neck | 33 mm |
Diameter (Metric) Inner | 33 mm |
For Use With (Application) | Storage and Shipment of Liquid Reagents |
Height (Metric) With Closure | 131 mm |
Material | HDPE |
No. per Case | 250 |
No. per Pack | 250 |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. |
Closure Included | Yes |
Mouth | Wide |
Shape | Round |
Height (Metric) | 131 mm |
Product Type | IP2 Bottle |
Height (English) | 5.1 in. |
Closure Material | Polypropylene |
Closure Size | 43-415 mm |
Diameter (Metric) | 61 mm |
Unit Size | Case of 250 |
Capacity | 32 oz., 1000 mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | White |
Description | IP2 Wide-Mouth Bottles |
Diameter (Metric) Inner Neck | 53 mm |
Diameter (Metric) Inner | 53 mm |
For Use With (Application) | Storage and Shipment of Liquid Reagents |
Height (Metric) With Closure | 199 mm |
Material | HDPE |
No. per Case | 50 |
No. per Pack | 50 |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. |
Closure Included | Yes |
Mouth | Wide |
Shape | Round |
Height (Metric) | 199 mm |
Product Type | IP2 Bottle |
Height (English) | 7.8 in. |
Closure Material | Polypropylene |
Closure Size | 63-415 mm |
Diameter (Metric) | 91 mm |
Unit Size | Case of 50 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | dimClosureSize | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
312199-0016 | Case of 125 | 500 mL | 647.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
312199-0004 | Case of 500 | 125 mL | 736.00 Online price 1,471.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
312199-0008 | Case of 250 | 250 mL | 1,009.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
312199-0032 | Case of 50 | 1000 mL | 470.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
Bottles and closures are bulk packed in separate polybags within the same carton
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.