Thermo Scientific Chemicals

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%(e.e.>99.5%), Thermo Scientific Chemicals

Catalog number: 312220010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%(e.e.>99.5%), Thermo Scientific Chemicals

Catalog number: 312220010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 312220010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20445-31-2
Appearance (Form)
Viscous liquid or low melting solid
Appearance (Color)
Clear colorless to light yellow
Titration with NaOH
98.5 to 101.5 %
Infrared spectrum
Conforms
Specific optical rotation
+71° to +74° (20°C, 589 nm) (c=2, CH3OH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text