Thermo Scientific Chemicals

Perfluoro-1-butanesulfonyl fluoride, 92%, Thermo Scientific Chemicals

Catalog number: 312341000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Perfluoro-1-butanesulfonyl fluoride, 92%, Thermo Scientific Chemicals

Catalog number: 312341000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 312341000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
375-72-4
IUPAC Name
nonafluorobutane-1-sulfonyl fluoride
Molecular Formula
C4F10O2S
InChI Key
LUYQYZLEHLTPBH-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(F)(=O)=O
Appearance (Color)
Clear colorless
GC
>=90 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text