Search Thermo Fisher Scientific
Scan 2D barcoded tubes into virtually any application with the compact Thermo Scientific™ VisionMate™ ST Single-Tube 2D Barcode Reader. Plug the unit into an available USB port and allow the drive to automatically install. Hold any Thermo Scientific™ 2D barcoded storage tube over the scanning window and decode the barcode instantly.
Description | VisionMate ST 2-D Bar Code Reader |
Quantity | 1/Pk., 1/Cs. |
Connection Type | USB |
Type | Barcode Reader |
Data Transfer Method | Comma-separated variable file, Microsoft™ Excel™ |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
3125 | Each | VisionMate ST 2-D Bar Code Reader | 2,569.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|