Thermo Scientific Chemicals

Triethylsilyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 312700100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Triethylsilyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 312700100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 312700100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79271-56-0
IUPAC Name
triethylsilyl trifluoromethanesulfonate
Molecular Formula
C7H15F3O3SSi
InChI Key
STMPXDBGVJZCEX-UHFFFAOYSA-N
SMILES
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
Appearance (Color)
Clear colorless to light brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration excess NaOH
>=98.5 %
Refractive index
1.3890 to 1.3920 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text