2,2,2-Trifluoroethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
2,2,2-Trifluoroethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trifluoroethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number 312780250
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS352-87-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless
Refractive index1.3600 to 1.3620 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only