2-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals
2-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 312930010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS312-94-7
IUPAC Name2-(trifluoromethyl)benzoyl chloride
Molecular FormulaC8H4ClF3O
InChI KeyMXIUWSYTQJLIKE-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4780 to 1.4810 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear yellow to pink
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only