Thermo Scientific Chemicals

3-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 312940050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 312940050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 312940050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2251-65-2
IUPAC Name
3-(trifluoromethyl)benzoyl chloride
Molecular Formula
C8H4ClF3O
InChI Key
RUJYJCANMOTJMO-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=CC(=C1)C(Cl)=O
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4760 to 1.4780 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text