3-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals
3-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 312940250
Price (USD)/ Each
174.00
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
174.00
Add to cart
3-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals
Catalog number312940250
Price (USD)/ Each
174.00
-
Add to cart
Chemical Identifiers
CAS2251-65-2
IUPAC Name3-(trifluoromethyl)benzoyl chloride
Molecular FormulaC8H4ClF3O
InChI KeyRUJYJCANMOTJMO-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4760 to 1.4780 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only