Thermo Scientific Chemicals

2-(Trifluoromethyl)acrylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 313100050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)acrylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 313100050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 313100050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
381-98-6
IUPAC Name
2-(trifluoromethyl)prop-2-enoate
Molecular Formula
C4H2F3O2
InChI Key
VLSRKCIBHNJFHA-UHFFFAOYSA-M
SMILES
[O-]C(=O)C(=C)C(F)(F)F
Infrared spectrum
Conforms
Melting point
48°C to 52°C
Titration with TBAH
>=97.5 %
Appearance (Color)
Transparent light yellow
Appearance (Form)
Chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text