Thermo Scientific Chemicals

3-Aminopropyltrimethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: 313251000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Aminopropyltrimethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: 313251000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 313251000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13822-56-5
IUPAC Name
(3-aminopropyl)trimethoxysilane
Molecular Formula
C6H17NO3Si
InChI Key
SJECZPVISLOESU-UHFFFAOYSA-N
SMILES
CO[Si](CCCN)(OC)OC
GC
>=94 %
Refractive index
1.4220 to 1.4250 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text