Thermo Scientific Chemicals

3-Cyanopropyldimethylchlorosilane, 95%, Thermo Scientific Chemicals

Catalog number: 313380500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Cyanopropyldimethylchlorosilane, 95%, Thermo Scientific Chemicals

Catalog number: 313380500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 313380500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18156-15-5
IUPAC Name
4-(chlorodimethylsilyl)butanenitrile
Molecular Formula
C6H12ClNSi
InChI Key
GPIARXZSVWTOMD-UHFFFAOYSA-N
SMILES
C[Si](C)(Cl)CCCC#N
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to straw yellow to brown
Refractive index
1.4450 to 1.4470 (20°C, 589 nm)
GC
>=94 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text