Thermo Scientific Chemicals

Octadecyltrimethoxysilane, 90%, Tech., Thermo Scientific Chemicals

Catalog number: 313601000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Octadecyltrimethoxysilane, 90%, Tech., Thermo Scientific Chemicals

Catalog number: 313601000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 313601000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3069-42-9
IUPAC Name
trimethoxy(octadecyl)silane
Molecular Formula
C21H46O3Si
InChI Key
SLYCYWCVSGPDFR-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC
Appearance (Color)
Clear to slightly hazy colorless to light yellow
Infrared spectrum
Conforms
GC
>=88 % Total octadecyltrimethoxysilanes
Appearance (Form)
Liquid
Appearance
May partially crystallize

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text