Thermo Scientific Chemicals

Ethoxytrimethylsilane, 95%, Thermo Scientific Chemicals

Catalog number: 313821000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethoxytrimethylsilane, 95%, Thermo Scientific Chemicals

Catalog number: 313821000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 313821000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1825-62-3
IUPAC Name
ethoxytrimethylsilane
Molecular Formula
C5H14OSi
InChI Key
RSIHJDGMBDPTIM-UHFFFAOYSA-N
SMILES
CCO[Si](C)(C)C
Refractive index
1.3730 to 1.3750 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text