Pierce™ Alkaline Phosphatase
Pierce™ Alkaline Phosphatase
Thermo Scientific™

Pierce™ Alkaline Phosphatase

This purified calf intestinal alkaline phosphatase (CIP) is supplied in Tris buffer and 50% glycerol for use in protein researchRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
3139120 mg
313925 x 20 mg
2 Options
Catalog number 31391
Price (USD)/ 20 mg
1,194.65
Online exclusive
1276.00 
Save 81.35 (6%)
-
Add to cart
Quantity:
20 mg
Request bulk or custom format
Price (USD)/ 20 mg
1,194.65
Online exclusive
1276.00 
Save 81.35 (6%)
Add to cart
Pierce™ Alkaline Phosphatase
Catalog number31391
Price (USD)/ 20 mg
1,194.65
Online exclusive
1276.00 
Save 81.35 (6%)
-
Add to cart
This purified calf intestinal alkaline phosphatase (CIP) is supplied in Tris buffer and 50% glycerol for use in protein research methods. The main applications for alkaline phosphatase in molecular biology and protein research are to remove 5'-phosphate groups from DNA or as a reporter system for immunoassays such as ELISA. For these latter methods, the enzyme is usually conjugated to specific primary or secondary antibodies and its activity is detected with a color-forming (or light-generating) substrate.

Features of Alkaline Phosphatase:

Purified form—ready to dilute and conjugate without prior dialysis
Concentrated—approximately 20 mg/mL (lot-specific value reported)
High specific activity—typically greater than 1600 units/mg (lot-specific value reported)
Tris buffered solution—supplied in 5 mM Tris, 5 mM magnesium chloride and 0.1 mM zinc chloride,
pH ∼7.0 in 50% glycerol

One unit equals the amount of protein required to hydrolyze one micromole of p-nitrophenyl phosphate (PNPP) per minute at 25°C in a glycine buffer, pH 9.6.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormSolid
For Use With (Application)ELISA, Immunohistochemistry
Product LinePierce™
Label or DyeAP (Alkaline Phosphatase)
Quantity20 mg
Unit Size20 mg
Contents & Storage
Store at 2–8°C.