Thermo Scientific Chemicals

chlorotripropylsilane, 98%, Thermo Scientific Chemicals

Catalog number: 313930250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

chlorotripropylsilane, 98%, Thermo Scientific Chemicals

Catalog number: 313930250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 313930250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
995-25-5
IUPAC Name
chlorotripropylsilane
Molecular Formula
C9H21ClSi
InChI Key
ACTAPAGNZPZLEF-UHFFFAOYSA-N
SMILES
CCC[Si](Cl)(CCC)CCC
Appearance (Color)
Clear colorless
Refractive index
1.4380 to 1.4420 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text