Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles, which include a sealing closure for added leakproof† assurance at maximum speeds when spinning hazardous materials. Molded of rigid and robust polypropylene copolymer, these translucent bottles are designed for pelleting applications, such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations. Additionally, these PPCO bottles are autoclavable and offer excellent mechanical strength.
Capacity (Metric) | 250 mL |
Capacity (English) | 8.45 oz. |
Color | Translucent |
Diameter (Metric) Outer Neck | 60.7 mm |
Material | PPCO |
Length (Metric) | 133.4 mm |
No. per Pack | 4 |
Nominal Capacity | 250 mL |
Closure Included | Yes |
Autoclavable | Autoclavable |
Graduated | No |
Max. Force | 27,500 x g |
Shape | Round |
Height (Metric) | 132.8 mm |
Product Type | Centrifuge Bottle |
Width (English) | 2.40 in. |
Length (English) | 5.23 in. |
Height (English) | 5.23 in. |
Closure Material | Polypropylene, red rubber |
Diameter (Metric) | 61.1 mm |
Closure Type | Sealing Cap Assembly with Silicone Gasket |
Through the Closure Dispensing | No |
Unit Size | Pack of 4 |
Capacity (Metric) | 250 mL |
Capacity (English) | 8.45 oz. |
Color | Translucent |
Diameter (Metric) Outer Neck | 60.7 mm |
Material | PPCO |
Length (Metric) | 133.4 mm |
No. per Pack | 4 |
Nominal Capacity | 250 mL |
Closure Included | Yes |
Autoclavable | Autoclavable |
Graduated | No |
Max. Force | 27,500 x g |
Shape | Round |
Height (Metric) | 132.8 mm |
Product Type | Centrifuge Bottle |
Width (English) | 2.40 in. |
Length (English) | 5.23 in. |
Height (English) | 5.23 in. |
Closure Material | Polypropylene, red rubber |
Diameter (Metric) | 61.1 mm |
Closure Type | Sealing Cap Assembly with Silicone Gasket |
Through the Closure Dispensing | No |
Unit Size | Case of 36 |
Capacity (Metric) | 450 mL |
Capacity (English) | 16.9 oz. |
Color | Translucent |
Material | PPCO |
Length (Metric) | 160 mm |
No. per Pack | 4 |
Nominal Capacity | 450 mL |
Closure Included | Yes |
Autoclavable | Autoclavable |
Graduated | No |
Max. Force | 13,700 x g |
Shape | Round |
Height (Metric) | 159.3 mm |
Product Type | Centrifuge Bottle |
Width (English) | 2.735 in. |
Length (English) | 2.735 in. |
Height (English) | 6.27 in. |
Closure Material | Polypropylene, red rubber |
Diameter (Metric) | 69.5 mm |
Closure Type | Sealing Cap Assembly with Silicone Gasket |
Through the Closure Dispensing | No |
Unit Size | Case of 24 |
Capacity (Metric) | 450 mL |
Capacity (English) | 16.9 oz. |
Color | Translucent |
Material | PPCO |
Length (Metric) | 160 mm |
No. per Pack | 4 |
Nominal Capacity | 450 mL |
Closure Included | Yes |
Autoclavable | Autoclavable |
Graduated | No |
Max. Force | 13,700 x g |
Shape | Round |
Height (Metric) | 159.3 mm |
Product Type | Centrifuge Bottle |
Width (English) | 2.735 in. |
Length (English) | 2.735 in. |
Height (English) | 6.27 in. |
Closure Material | Polypropylene, red rubber |
Diameter (Metric) | 69.5 mm |
Closure Type | Sealing Cap Assembly with Silicone Gasket |
Through the Closure Dispensing | No |
Unit Size | Pack of 4 |
Catalog Number | Specifications | Unit Size | Nominal Capacity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
3141-0250PK | Pack of 4 | 250 mL | 198.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3141-0250 | Case of 36 | 250 mL | 1,249.21 Online price 1,404.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3141-0500 | Case of 24 | 450 mL | 1,236.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3141-0500PK | Pack of 4 | 450 mL | 261.70 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Requires:
When spinning 250mL bottle in a Sorvall GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required.
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.