Thermo Scientific Chemicals

3,5-Dichlorophenyl isocyanate, 96%, Thermo Scientific Chemicals

Catalog number: 314140010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Dichlorophenyl isocyanate, 96%, Thermo Scientific Chemicals

Catalog number: 314140010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 314140010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34893-92-0
IUPAC Name
1,3-dichloro-5-isocyanatobenzene
Molecular Formula
C7H3Cl2NO
InChI Key
XEFUJGURFLOFAN-UHFFFAOYSA-N
SMILES
ClC1=CC(=CC(Cl)=C1)N=C=O
GC
>=95 %
Appearance (Color)
White to light yellow
Appearance (Form)
Low melting solid
Infrared spectrum
Conforms
Melting point
29°C to 35°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text