Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 314170050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 314170050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 314170050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16588-74-2
IUPAC Name
1-isocyanato-3,5-bis(trifluoromethyl)benzene
Molecular Formula
C9H3F6NO
InChI Key
NRSSOFNMWSJECS-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC(=C1)N=C=O)C(F)(F)F
Appearance (Form)
Liquid
GC
>=97.5 %
Refractive index
1.4290 to 1.4330 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text