Thermo Scientific Chemicals

Benzoyl isocyanate, 90%, Tech., Thermo Scientific Chemicals

Catalog number: 314280010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Benzoyl isocyanate, 90%, Tech., Thermo Scientific Chemicals

Catalog number: 314280010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 314280010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4461-33-0
IUPAC Name
benzoyl isocyanate
Molecular Formula
C8H5NO2
InChI Key
LURYMYITPCOQAU-UHFFFAOYSA-N
SMILES
O=C=NC(=O)C1=CC=CC=C1
Appearance (Color)
Clear to hazy colorless to yellow
Appearance (Form)
Liquid or solid
Infrared spectrum
Conforms
GC
>=88 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text