Thermo Scientific Chemicals

Ethoxycarbonyl isocyanate, 90%, tech., Thermo Scientific Chemicals

Catalog number: 314310010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethoxycarbonyl isocyanate, 90%, tech., Thermo Scientific Chemicals

Catalog number: 314310010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 314310010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19617-43-7
IUPAC Name
ethyl N-carbonylcarbamate
Molecular Formula
C4H5NO3
InChI Key
VMVZGGPZNHFGKS-UHFFFAOYSA-N
SMILES
CCOC(=O)N=C=O
GC
>=88 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to slightly colored

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text