Ethoxycarbonyl isocyanate, 90%, tech., Thermo Scientific Chemicals
Ethoxycarbonyl isocyanate, 90%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethoxycarbonyl isocyanate, 90%, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 314310250
Price (USD)/ Each
1,881.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
1,881.00
Add to cart
Ethoxycarbonyl isocyanate, 90%, tech., Thermo Scientific Chemicals
Catalog number314310250
Price (USD)/ Each
1,881.00
-
Add to cart
Chemical Identifiers
CAS19617-43-7
IUPAC Nameethyl N-carbonylcarbamate
Molecular FormulaC4H5NO3
InChI KeyVMVZGGPZNHFGKS-UHFFFAOYSA-N
SMILESCCOC(=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=88 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to slightly colored
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only