Thermo Scientific Chemicals

1,4-Diisocyanatobutane, 97%, Thermo Scientific Chemicals

Catalog number: 314380250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1,4-Diisocyanatobutane, 97%, Thermo Scientific Chemicals

Catalog number: 314380250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 314380250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4538-37-8
IUPAC Name
1,4-diisocyanatobutane
Molecular Formula
C6H8N2O2
InChI Key
OVBFMUAFNIIQAL-UHFFFAOYSA-N
SMILES
O=C=NCCCCN=C=O
Refractive index
1.4510 to 1.4530 (20°C, 589 nm)
Appearance (Color)
Clear light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text