3-(chloromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals
3-(chloromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(chloromethyl)benzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 314430050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-(chloromethyl)benzoyl chloride
Molecular FormulaC8H6Cl2O
InChI KeyYCAIYRWHKSJKEB-UHFFFAOYSA-N
SMILESClCC1=CC=CC(=C1)C(Cl)=O
Molecular Weight (g/mol)189.04
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.5740 to 1.5760 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only