1-Chloroethyl chloroformate, 97+%, Thermo Scientific Chemicals
1-Chloroethyl chloroformate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Chloroethyl chloroformate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 314460250
Price (USD)/ Each
90.65
Online exclusive
106.00 
Save 15.35 (14%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Online exclusive
106.00 
Save 15.35 (14%)
Add to cart
1-Chloroethyl chloroformate, 97+%, Thermo Scientific Chemicals
Catalog number314460250
Price (USD)/ Each
90.65
Online exclusive
106.00 
Save 15.35 (14%)
-
Add to cart
Chemical Identifiers
CAS50893-53-3
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.4220 to 1.4240 (20°C, 589 nm)
GC>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only