Thermo Scientific Chemicals

1-Chloro-2-methylpropyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog number: 314470010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Chloro-2-methylpropyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog number: 314470010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 314470010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92600-11-8
IUPAC Name
(1R)-1-chloro-2-methylpropyl carbonochloridate
Molecular Formula
C5H8Cl2O2
InChI Key
BOGLZTPABVHWDD-BYPYZUCNSA-N
SMILES
CC(C)[C@@H](Cl)OC(Cl)=O
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Refractive index
1.4310 to 1.4330 (20°C, 589 nm)
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text