1-Chloro-2-methylpropyl chloroformate, 98%, Thermo Scientific Chemicals
1-Chloro-2-methylpropyl chloroformate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Chloro-2-methylpropyl chloroformate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 314470050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS92600-11-8
IUPAC Name(1R)-1-chloro-2-methylpropyl carbonochloridate
Molecular FormulaC5H8Cl2O2
InChI KeyBOGLZTPABVHWDD-BYPYZUCNSA-N
SMILESCC(C)[C@@H](Cl)OC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.4310 to 1.4330 (20°C, 589 nm)
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only