Octyl chloroformate, 97%, Thermo Scientific Chemicals
Octyl chloroformate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Octyl chloroformate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 314550250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7452-59-7
IUPAC Nameoctyl carbonochloridate
Molecular FormulaC9H17ClO2
InChI KeyVFXVAXFIFHSGNR-UHFFFAOYSA-N
SMILESCCCCCCCCOC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Assay>=96.0 %
Refractive index1.4300 to 1.4320 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only