Thermo Scientific Chemicals

N-Methylpropylamine, 96%, Thermo Scientific Chemicals

Catalog number: 314690050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Methylpropylamine, 96%, Thermo Scientific Chemicals

Catalog number: 314690050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 314690050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
627-35-0
IUPAC Name
methyl(propyl)amine
Molecular Formula
C4H11N
InChI Key
GVWISOJSERXQBM-UHFFFAOYSA-N
SMILES
CCCNC
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
GC
>=95 %
Infrared spectrum
Conforms
Refractive index
1.3890 to 1.3940 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text